missing translation for 'onlineSavingsMsg'
Learn More

DLL3 Antibody (7B7), Novus Biologicals™

Product Code. 30043448 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
50 μL
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30043448 50 μL 50µL
30043783 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30043448 Supplier Novus Biologicals Supplier No. NBP32661850ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

DLL3 Monoclonal antibody specifically detects DLL3 in Human samples. It is validated for Flow Cytometry, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen DLL3
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 7B7
Conjugate Unconjugated
Dilution Flow Cytometry 1:50-1:200, ELISA
Formulation 50% Glycerol, 0.01M PBS, PH 7.4
Gene Alias delta (Drosophila)-like 3, Delta3, delta-like 3 (Drosophila), delta-like protein 3, Drosophila Delta homolog 3, SCDO1delta3
Host Species Human
Immunogen Recombinant Human DLL3 protein [UniProt Q9NYJ7]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 10683
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.