missing translation for 'onlineSavingsMsg'
Learn More

DL-Thioctic acid, 98-99%, MP Biomedicals™

Product Code. 11398739 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398739 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11398739 Supplier MP Biomedicals Supplier No. 0210113825
  • ONLINE EXCLUSIVE PRICE
    340.00 EUR
    323.40 EUR / 25g
    Save 16.60 EUR
    4% Off
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1077-28-7
Assay Percent Range 98 to 99%
Molecular Formula C8H14O2S2
MDL Number MFCD00005474
Quantity 25 g
Synonym α-Lipoic acid
InChI Key AGBQKNBQESQNJD-UHFFFAOYNA-N
SMILES OC(=O)CCCCC1CCSS1
IUPAC Name 5-(1,2-dithiolan-3-yl)pentanoic acid
Molecular Weight (g/mol) 206.32
Formula Weight 206.3
Grade Cell Culture Reagent
Physical Form Crystalline Powder
Chemical Name or Material DL-Thioctic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.