Learn More
DL-Menthyl Stearate, TRC
Specifications
Specifications
| Chemical Name or Material | DL-Menthyl Stearate |
| CAS | 1014622-89-9 |
| Molecular Formula | C28H54O2 |
| Quantity | 500 mg |
| Synonym | rel-(1R,2S,5R)-5-Methyl-2-(1-methylethyl)cyclohexyl Octadecanoate |
| InChI Formula | InChI=1S/C28H54O2/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-28(29)30-27-23-25(4)21-22-26(27)24(2)3/h24-27H,5-23H2,1-4H3/t25-,26+,27-/m1/s1 |
| SMILES | C(C)(C)[C@H]1[C@H](OC(CCCCCCCCCCCCCCCCC)=O)C[C@H](C)CC1 |
| IUPAC Name | (1R,2S,5R)-2-isopropyl-5-methylcyclohexyl stearate |
| Molecular Weight (g/mol) | 422.73 |
| Formula Weight | 422.41 |