missing translation for 'onlineSavingsMsg'
Learn More

dl-Malic Acid, Millipore™

Product Code. 10794041
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10794041 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10794041 Supplier Millipore Supplier No. 1003820250
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For biochemistry

TRUSTED_SUSTAINABILITY

Specifications

CAS 6915-15-7
Melting Point 128°C to 132°C
Density 1.60 g/cm3 at 20°C
Quantity 250 g
MDL Number MFCD00064212
Synonym DL-Malic acid, DL-Hydroxysuccinic acid, Hydroxybutanedioic acid, DL-Malic acid
Molecular Weight (g/mol) 134.09
Chemical Name or Material DL-Malic Acid
compliance-icons
Product Identifier
  • DL-Malic acid for biochemistry
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: DL-malic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.