missing translation for 'onlineSavingsMsg'
Learn More

DL-Epinephrine, MP Biomedicals™

Product Code. 11496842 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496842 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11496842 Supplier MP Biomedicals Supplier No. 0215106425
  • ONLINE EXCLUSIVE PRICE
    393.00 EUR
    373.80 EUR / 25g
    Save 19.20 EUR
    4% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 329-65-7
Color Tan to White
Molecular Formula C9H13NO3
Quantity 25 g
Synonym dl-adrenaline, racepinephrine, racepinefrine, dl-epinephrine, +--adrenaline, epinephrine racemic, racepinefrina, racepinefrinum, +--epinephrine, 4-1-hydroxy-2-methylamino ethyl benzene-1,2-diol
InChI Key UCTWMZQNUQWSLP-UHFFFAOYSA-N
SMILES CNCC(C1=CC(=C(C=C1)O)O)O
IUPAC Name 4-[1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
Molecular Weight (g/mol) 183.207
PubChem CID 838
ChEBI CHEBI:33568
Formula Weight 183.21
Physical Form Crystalline Powder
Chemical Name or Material DL-Epinephrine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.