missing translation for 'onlineSavingsMsg'
Learn More

DL-Dithiothreitol, MP Biomedicals™

Product Code. 11437690 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11437690 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11437690 Supplier MP Biomedicals Supplier No. 02100597.2
  • ONLINE EXCLUSIVE PRICE
    44.10 EUR
    42.00 EUR / 250mg
    Save 2.10 EUR
    4% Off
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Known as Clelands reagent. DL-Dithiothreitol, MP Biomedicals is a protective agent that quantitatively reduces disulfides (-S-S- to -SH). In this reaction, DTT is oxidized to the cyclic disulfide, ensuring the reduction of other disulfides in solution. Di

Dithiothreitol is useful for stabilizing sulfhydryl containing enzymes. Effective in sample buffers for reducing protein disulfide bonds prior to SDS-PAGE. DTT can also be used for reducing the disulfide bridge of the cross-linker N,N'-bis(acryloyl)cystamine to break apart the matrix of a polyacrylamide gel. Typically, a seven fold lower concentration of DTT (100 mM) is needed than is used for 2-mercaptoethanol (5% v/v, 700 mM).

DTT has been used:

  • As one of the reactants in the reduction and alkylation of αs1-Casein, the major allergen of cow′s milk
  • As a component of medium for the demembranation and reactivation of spermatozoa
  • To maintain stability of the enzyme as thiol effectively protects the active sites of the biocatalyst
  • As a reducing agent to test the specificity of the reaction of N-Ethylmaleimide with sulfhydryl groups

Specifications

CAS 12-3-3483
Melting Point 40°C to 50°C (literature)
pH 4.0 to 6.0
Molecular Formula C4H10O2S2
Quantity 250 mg
Synonym dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt
Solubility Information Soluble in water (50mg/mL, clear and colorless solution); soluble in ethanol, acetone, ethylate, chloroform and ether.
InChI Key VHJLVAABSRFDPM-IMJSIDKUSA-N
SMILES C(C(C(CS)O)O)S
IUPAC Name (2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol
Molecular Weight (g/mol) 154.24
PubChem CID 446094
ChEBI CHEBI:42170
Formula Weight 154.2
Percent Purity ≥99.0%
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.