Learn More
dl-Carnitine Hydrochloride, Sigma-Aldrich™
Product Code. 15862331
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15862331 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15862331 Supplier Sigma Aldrich Supplier No. 8417740025
 Restricted Item
Estimated Shipment: 24-11-2026
MQ200 Graded - For Research and Non-regulated Industrial Applications
For synthesis
TRUSTED_SUSTAINABILITY
Specifications
CAS 461-05-2
MDL Number MFCD00011904
Quantity 25 g
Synonym DL-Carnitine hydrochloride, DL-Carnitine hydrochloride
Molecular Weight (g/mol) 197.66
Grade Research