missing translation for 'onlineSavingsMsg'
Learn More

DL-3-phenyllactic Acid-d3, TRC

Product Code. 17939089
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17939089 5 mg 5mg
17929089 10 mg 10mg
17938909 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17939089 Supplier Toronto Research Chemicals Supplier No. TRCP3355675MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 490034-49-6
Molecular Formula C9H7D3O3
Molecular Weight (g/mol) 169.19
Synonym Benzenepropanoic-α,β,β-d3 acid, α-hydroxy- (9CI), α-Hydroxybenzenepropanoic-α,β,β-d3 acid, 2-Hydroxy-3-phenylpropanoic-2,3,3-d3 acid, 3-Phenyllactic-2,3,3-d3 acid, DL-3-Phenyllactic-2,3,3-d3 acid, DL-3-Phenyllactic-2,3,3-D3 acid, β-Phenyllactic-α,β,β-d3 acid, DL-3-Phenyllactic acid-d3
SMILES [2H]C(C(C(O)=O)(O)[2H])([2H])C1=CC=CC=C1

Specifications

Quantity 5 mg
Formula Weight 169.0818
Percent Purity >95%
Chemical Name or Material DL-3-phenyllactic Acid-d3
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.