Learn More
Dixyrazine, MedChemExpress
Product Code. 16473189
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Förpackningsstorlek:
10mg
1mg
20mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity unitSize
16473189 1 mg 1mg
16432408 5 mg 5mg
16430209 10 mg 10mg
16410726 20 mg 20mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16473189 Leverantör Medchem Express Leverantörsnummer HYU00153
This item requires a price update prior to being ordered.
Request a Price for this item here.
Denna artikel kan inte returneras. Se returpolicy
MedChemExpress Dixyrazine, a phenothiazine derivative, can prevent brain oedema induced by intracarotid injection of protamine sulphate.
Dixyrazine, a phenothiazine derivative, can prevent brain oedema induced by intracarotid injection of protamine sulphate.
Specifikationer
Chemical Name or Material Dixyrazine
CAS 2470-73-7
Color White
Purity Grade Notes Research
Molecular Formula C24H33N3O2S
Quantity 1 mg
Solubility Information DMSO : 100 mg/mL (233.86 mM; Need ultrasonic)
SMILES CC(CN1C2=C(C=CC=C2)SC3=CC=CC=C13)CN4CCN(CCOCCO)CC4
Molecular Weight (g/mol) 427.6
Formula Weight 427.6
Percent Purity 99.14%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Visa mer Visa mindre
Research purposes only
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.