missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dityrosine Antibody (7D4), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
188.00 EUR - 588.00 EUR
Specifications
| Antigen | Dityrosine |
|---|---|
| Clone | 7D4 |
| Dilution | Western Blot 1:1000, ELISA 1:1000, Immunocytochemistry/Immunofluorescence 1:50 |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18248052
|
Novus Biologicals
NBP2-59360 |
100 μg |
588.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18661578
|
Novus Biologicals
NBP2-59360-25ug |
25 μg |
188.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dityrosine Monoclonal specifically detects Dityrosine in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Dityrosine | |
| Western Blot 1:1000, ELISA 1:1000, Immunocytochemistry/Immunofluorescence 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| IgG1 | |
| Protein G purified |
| 7D4 | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Synthetic Dityrosine conjugated to Keyhole Limpet Hemocyanin (KLH). | |
| Primary | |
| Specific for dityrosine modified proteins. Does not cross-react with 3,5-dibromotyrosine or bromotyrosine modified proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title