missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dityrosine Antibody (7D4), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-59360
This item is not returnable.
View return policy
Description
Dityrosine Monoclonal specifically detects Dityrosine in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Dityrosine | |
| Monoclonal | |
| Unconjugated | |
| PBS pH 7.4, 50% glycerol with 0.09% Sodium Azide | |
| Synthetic Dityrosine conjugated to Keyhole Limpet Hemocyanin (KLH). | |
| 100 μg | |
| Primary | |
| Non-species specific | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 7D4 | |
| Western Blot 1:1000, ELISA 1:1000, Immunocytochemistry/Immunofluorescence 1:50 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Specific for dityrosine modified proteins. Does not cross-react with 3,5-dibromotyrosine or bromotyrosine modified proteins. | |
| Store at -20C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction