Learn More
Lenz Laborglasinstrumente™ Distilling Links
Product Code. 10234341
Change view
Click to view available options
Standard Taper Joint:
NS 14/23
NS 14/23, NS 14/23
NS 14/23, NS 29/32
NS 24/29
NS 29/32
NS 29/32, GL 14
NS 29/32, NS 14/23
NS 29/32, NS 29/32
Length (Metric) Jacket:
160 mm
250 mm
300 mm
350 mm
400 mm
Unit Size:
Each
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint Length (Metric) Jacket unitSize
10234341 NS 24/29 300 mm Each
10686755 NS 14/23 160 mm Each
10702682 NS 14/23, NS 14/23 160 mm Each
10147044 NS 29/32, GL 14 250 mm Each
10491865 NS 29/32 250 mm Each
10548081 NS 14/23, NS 29/32 250 mm Each
10634271 NS 29/32, NS 14/23 250 mm Each
10363181 NS 29/32, NS 29/32 350 mm Each
10376021 NS 14/23, NS 29/32 350 mm Each
11816214 NS 29/32 350 mm Each
10545670 NS 29/32 400 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 10234341 Supplier Lenz Laborglasinstrumente™ Supplier No. 05040003
Add to basket
Add to basket
Distilling Links for the connection of distillation thermometers of 55 mm built-in length.
  • With Liebig condenser
  • Outlet with NS cone
  • Water connection with glass olives, with NS socket
Specifications
Type Distilling Link
Material Duran Glass
Standard Taper Joint NS 24/29
Length (Metric) Jacket 300 mm