Learn More
Hanna Instruments™ Dissolved oxygen probe with 10m cable Hanna
Product Code. 10509120
Change view
Click to view available options
Length (Metric) Cable:
10 m
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Cable Unit Size
10509120 10 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10509120 Supplier Hanna Instruments™ Supplier No. HI76407/10
  • ONLINE EXCLUSIVE PRICE
    1027.95 EUR
    535.64 EUR / Pack of 1
    Save 492.31 EUR
    47% Off
 Restricted Item
Estimated Shipment: 27-10-2026