Learn More
Fisherbrand™ Dissecting Scissors
Product Code. 15277168
Change view
Click to view available options
Blade Style:
Curved
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Blade Style Unit Size
15277168 Curved Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Product Code. 15277168 Supplier Fisherbrand™ Supplier No. E108/01
Estimated Shipment: 21-10-2026
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Use these scissors for a variety of applications. These dissecting scissors are suitable for theatre, ward, veterinary, dental and chiropody use.
  • Stainless steel dissecting scissors
  • Closed shanks, fine point
  • Comply with the essential requirements of the Medical Devices Directive 93/42/EEC
  • Full traceability maintained throughout the manufacturing process.
  • Made from only high grade stainless steel in accordance to the specifications of ISO 7153-1
  • 10 year guarantee
Specifications
Product Type Dissecting Scissors
Blade Style Curved
Length (Metric) 105 mm
Material Stainless Steel