Learn More
Karl Hecht™ Disposable Volume End-to-End Capillaries
Product Code. 10093721
Change view
Click to view available options
Volume (Metric):
2 μL
10 μL
Unit Size:
Pack of 250
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10093721 2 μL Pack of 250
10513842 10 μL Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10093721 Supplier Karl Hecht™ Supplier No. 40551006
  • ONLINE EXCLUSIVE PRICE
    306.72 EUR
    27.00 EUR / Pack of 250
    Save 279.72 EUR
    91% Off
Estimated Shipment: 29-12-2026
Filling and discharging can be effected with or without a capillary holder.
  • Calibrated to contain from end to end
  • Conformity certified, CE, DIN ISO 7550
  • With capillary holder no. 40553001 (short tube with rubber teat)
  • Accuracy of ≤ ±0.5%
  • Precision of ≤ 1%
  • 250 pieces per pack, 4 packs per carton
Specifications
For Use With (Application) Pipetting
Includes Capillary holder no. 40553001
Packaging Type 250 per pack; 4 packs per carton
Volume (Metric) 2 μL