Learn More
Epredia™ Disposable Blade Carriers
Product Code. 12033119 Shop All Brand Products
Change view
Click to view available options
Product Type:
Blade Carrier E
Disposable Blade Carrier E
For Use With (Equipment):
Dovetail Style Unit
HM 310, HM 315 and HM 325
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type For Use With (Equipment) Unit Size
12033119 Blade Carrier E HM 310, HM 315 and HM 325 Each
10680032 Disposable Blade Carrier E Dovetail Style Unit Each
12647876 Disposable Blade Carrier E Dovetail Style Unit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12033119 Supplier Epredia™ Supplier No. 705800
  • ONLINE EXCLUSIVE PRICE
    1830.00 EUR
    1458.66 EUR / Pack of 1
    Save 371.34 EUR
    20% Off
In Stock
Utilize low- and high-profile blades with the quick clamp system of Epredia™ Disposable Blade Carriers.
Disposable Blade Carriers
  • Blade angle and blade adjustment levers are removable to help protect user settings (Version 2 and 3 units only)
  • Concentric lever locks onto the dovetail to help reduce accidental movement of the holder (Version 2 and 3 units only)
  • Quick clamp system for high- and low-profile blades
  • Adjustable clearance angle
  • Moveable knife guard covers the blade entirely in any position
  • Knife carrier base included
TRUSTED_SUSTAINABILITY
Specifications
Description Blade carrier E
Product Type Blade Carrier E
For Use With (Equipment) HM 310, HM 315 and HM 325