Learn More
Epredia™ Disposable Blade Carriers
Product Code. 10680032 Shop All Brand Products
Change view
Click to view available options
Product Type:
Blade Carrier E
Disposable Blade Carrier E
For Use With (Equipment):
Dovetail Style Unit
HM 310, HM 315 and HM 325
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type For Use With (Equipment) Unit Size
10680032 Disposable Blade Carrier E Dovetail Style Unit Each
12647876 Disposable Blade Carrier E Dovetail Style Unit Each
12033119 Blade Carrier E HM 310, HM 315 and HM 325 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10680032 Supplier Epredia™ Supplier No. 705050
  • ONLINE EXCLUSIVE PRICE
    75.25 EUR
    70.19 EUR / Pack of 1
    Save 5.06 EUR
    6% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Utilize low- and high-profile blades with the quick clamp system of Epredia™ Disposable Blade Carriers.
Disposable Blade Carriers
  • Blade angle and blade adjustment levers are removable to help protect user settings (Version 2 and 3 units only)
  • Concentric lever locks onto the dovetail to help reduce accidental movement of the holder (Version 2 and 3 units only)
  • Quick clamp system for high- and low-profile blades
  • Adjustable clearance angle
  • Moveable knife guard covers the blade entirely in any position
  • Knife carrier base included
TRUSTED_SUSTAINABILITY
Specifications
Description E Type blade carrier for dovetail style units
Product Type Disposable Blade Carrier E
For Use With (Equipment) Dovetail Style Unit