Learn More
CellPath Dispocut™ Disposable Cut-Up Boards
Product Code. 12806653
Change view
Click to view available options
Dimensions (L x W):
297 x 420 mm
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
12806653 297 x 420 mm Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12806653 Supplier CellPath Supplier No. CGB030251A
  • ONLINE EXCLUSIVE PRICE
    121.84 EUR
    96.41 EUR / Pack of 12
    Save 25.43 EUR
    20% Off
 Restricted Item
Estimated Shipment: 04-12-2026
For routine and high risk specimens
  • Disposable by incineration
  • Manufactured from 3mm, three ply foam filled board for strength and rigidity
  • High degree of moisture resistance
Specifications
Product Type Dispocut Board White
Color White
Disposable Single-use
For Use With (Application) Routine and high risk specimens
Dimensions (L x W) 297 x 420 mm