Learn More
BRAND™ Dispensing Cylinder with Valve Block
Product Code. 15693987 Shop All Brand Products
Change view
Click to view available options
For Use With (Equipment):
10 mL Burette
50 mL Burette
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15693987 10 mL Burette Each
11663219 50 mL Burette Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15693987 Supplier BRAND™ Supplier No. 707533
  • ONLINE EXCLUSIVE PRICE
    107.00 EUR
    96.08 EUR / Pack of 1
    Save 10.92 EUR
    10% Off
Estimated Shipment: 08-12-2026
For use with Titrette™ Bottletop Burette. Accommodates all serial number ranges.
  • For use with Titrette™ Bottletop Burette
  • Fits all serial number ranges
  • *For 50mL instruments with serial numbers beginning with 01-12 AND letters A-J, must order both Dispensing cylinder and Titrating tube as a pair.
TRUSTED_SUSTAINABILITY
Specifications
Type Dispensing Cylinder
Includes Valve head for titrette
For Use With (Equipment) 10 mL Burette