Learn More
Fisherbrand™ Benchtop Double Dispensing Bin
Product Code. 11304065 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Color:
Neon Green
Dimensions (L x W x H):
180 x 155 x 165 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Dimensions (L x W x H) Unit Size
11304065 Neon Green 180 x 155 x 165 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11304065
  • ONLINE EXCLUSIVE PRICE
    76.55 EUR
    68.19 EUR / Pack of 1
    Save 8.36 EUR
    10% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket
Access your lab materials with ease, even with gloved hands. Fisherbrand™ Benchtop Double Dispensing Bin is ergonomically designed with a large, angled opening and a lid that stays open when tilted.
Easily access your lab materials, even with gloved hands! Ideal for holding many different laboratory items including micro-centrifuge tubes.
Ergonomically designed
  • Large, angled openings
  • Oversized lip
  • Lid remains open when tilted back
  • Includes a removable compartment divider
Perfect for benchtop and wall mounted use
  • Nonskid rubber feet
  • Pre-drilled holes and included screws for wall mounting
Specifications
Type Dispensing Bin
Material Acrylic
Color Neon Green
Quantity 1 Each
Dimensions (L x W x H) 180 x 155 x 165 mm
Length (Metric) 180 mm
Width (Metric) 155 mm
Height (Metric) 165 mm