Learn More
Fisherbrand™ Dispenser tips
Product Code. 15173646 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
0.1 mL
0.5 mL
1.25 mL
2.5 mL
5 mL
5.0 mL
12.5 mL
25 mL
50 mL
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Pack of 25
Pack of 50
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Sterility unitSize
15173646 2.5 mL Non-sterile Pack of 100
15163646 0.1 mL Non-sterile Pack of 100
15133646 0.5 mL Non-sterile Pack of 100
15153646 1.25 mL Non-sterile Pack of 100
15143646 5.0 mL Non-sterile Pack of 100
15193646 12.5 mL Non-sterile Pack of 100
15183646 25 mL Non-sterile Pack of 50
15103656 50 mL Non-sterile Pack of 25
15113656 0.1 mL Sterile Pack of 100
15123656 0.5 mL Sterile Pack of 100
15143656 1.25 mL Sterile Pack of 100
15133656 2.5 mL Sterile Pack of 100
15153656 5 mL Sterile Pack of 100
15173656 12.5 mL Sterile Pack of 100
15183656 50 mL Sterile Pack of 25
15163656 25 mL Sterile Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Product Code. 15173646
  • ONLINE EXCLUSIVE PRICE
    77.30 EUR
    69.85 EUR / Pack of 100
    Save 7.45 EUR
    9% Off
View all promotions
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
Robust, stable, and durable dispenser tips for several brands of pipettors. For dispensing of fluids with high viscosity and high vapor pressure.
  • These disposable tips are suitable for the following multiple dispensers:
  • BRAND Handystep™ S, HandyStep™ electronic, HandyStep™ touch/ S
  • Eppendorf Multipette™ 4780
  • Gilson Distriman™ and Gilson Repetman™
  • Ritter Ripette™ pro
  • EDOS™ 5221
  • Rainin AutoRep™ E, Rainin AutoRep™ S, Rainin AutoRep™ M
  • STARLAB RepeatOne™
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 2.5 mL
Sterility Non-sterile
Certifications/Compliance ISO 8655
For Use With (Application) For dispensing of fluids with high viscosity, high vapor pressure
Product Type Dispenser Tip