missing translation for 'onlineSavingsMsg'
Learn More

Disodium Zinc Ethylenediaminetetraacetate Tetrahydrate, TRC

Product Code. 30179916
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179916 100 g 100g
30179792 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30179916 Supplier Toronto Research Chemicals Supplier No. TRCD493658100G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 176736-49-5
Molecular Formula C10H12N2O8Zn • 2Na • 4H2O
Molecular Weight (g/mol) 471.65
Synonym Sodium (OC-6-21)-[[N,N'-1,2-Ethanediylbis[N-[(carboxy-κO)methyl]glycinato-κN,κO]](4-)]zincate(2-) Hydrate;
IUPAC Name Zincate(2-), [[N,N′-1,2-ethanediylbis[N-[(carboxy-κO)methyl]glycinato-κN,κO]](4-)]-, sodium, hydrate (1:2:4)
SMILES O=C1[O-][Zn+2]2([O-]C3=O)([N](C1)(C3)CC[N]2(C4)CC5=O)([O-]5)[O-]C4=O

Specifications

Quantity 100 g
InChI Formula InChI=1S/C10H16N2O8.2Na.4H2O.Zn/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;4*1H2;/q;2*+1;;;;;+2/p-4
Formula Weight 470.01
Chemical Name or Material Disodium Zinc Ethylenediaminetetraacetate Tetrahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.