Learn More
Veolia Water Systems™ Disinfection Tablets
Product Code. 12826753
Change view
Click to view available options
Type:
Disinfection Tablets
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
12826753 Disinfection Tablets Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12826753 Supplier Veolia Water Systems™ Supplier No. CT3
 Restricted Item
Estimated Shipment: 23-12-2026
Use Disinfection Tablets to sanitise tubing, or pipework and system parts that come into contact with water, to remove bacteria, reduce bio film and protect the system against bacterial growth.
  • For regular disinfection of system parts that come into contact with water
Specifications
Description Effervescent tablets
Type Disinfection Tablets
For Use With (Application) Sanitising Tubing and Pipework
Weight (Metric) 0.99 kg