Learn More
Johnson Johnson Medical™ Disinfectant granules Presept
Description
Reacts with water in body fluid spillages to produce a rapid acting wide spectrum disinfectant effective against bacteria, fungi, viruses (including AIDS virus) and bacterial spores.
Effervescent, containing sodium dichloroisocyanurate (NaDCC). The granules are applied over the spillage, left for two minutes and removed for disposal. The surface is then wiped with a damp cloth, which is disposed of along with used granules. Should not be mixed with acids or cationic detergents.Use biocides safely. Always read the label and product information before use.
Granules contain and solidify the spillage, enabling easy removal, and preventing the spread of contamination.
Effervescent, containing sodium dichloroisocyanurate (NaDCC). The granules are applied over the spillage, left for two minutes and removed for disposal. The surface is then wiped with a damp cloth, which is disposed of along with used granules. Should not be mixed with acids or cationic detergents.Use biocides safely. Always read the label and product information before use.
Granules contain and solidify the spillage, enabling easy removal, and preventing the spread of contamination.