Learn More
BRAND™ Discharge Valves for Dispensette™ S Bottletop Dispensers
Product Code. 15643977 Shop All Brand Products
Change view
Click to view available options
For Use With (Equipment):
Dispensette S ≥25 mL
Dispensette S ≤10 mL
Dispensette S Organic, 25, 50, 100 mL
Dispensette S Organic, 5 and 10 mL
Dispensette S Trace Analysis Pt-Ir
Dispensette S Trace Analysis Ta
Dispensette S, 1 and 2mL
Autoclavable:
Autoclavable
Not Autoclavable
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Autoclavable Unit Size
15643977 Dispensette S ≤10 mL Autoclavable Each
15663977 Dispensette S Organic, 25, 50, 100 mL Autoclavable Each
15523583 Dispensette S ≥25 mL Autoclavable Each
15653977 Dispensette S Organic, 5 and 10 mL Autoclavable Each
15673977 Dispensette S Trace Analysis Pt-Ir Not Autoclavable Each
15683977 Dispensette S Trace Analysis Ta Not Autoclavable Each
15603987 Dispensette S, 1 and 2mL Autoclavable Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 15643977 Supplier BRAND™ Supplier No. 6727
  • ONLINE EXCLUSIVE PRICE
    35.70 EUR
    33.69 EUR / Pack of 1
    Save 2.01 EUR
    5% Off
View all promotions
Estimated Shipment: 15-10-2026
Valves for Dispensette™ S / S Organic / S Trace Analysis
  • Discharge valves are manufactured of PFA/Boro 3.3/ceramic components. The Dispensette™ S discharge valve has a platinum-iridium valve spring; the Dispensette™ S Organic discharge valve has a tantalum valve spring.
  • Discharge valves for the Dispensette™ S Trace Analysis are available with either a platinum-iridium or tantalum valve spring for specific applications (e.g., Pt-Ir valve for dispensing HF). Available in two or three sizes: one for 1mL and 2mL models; one for 5mL and 10mL models; one for 25mL, 50mL and 100mL models
TRUSTED_SUSTAINABILITY
Specifications
Product Type Discharge Valve
Autoclavable Autoclavable
Color Translucent
Material ETFE, Borosilicate Glass, Ceramic
For Use With (Application) Dispensing liquid
For Use With (Equipment) Dispensette S ≤10 mL
Includes Discharge Valve Only