Learn More
BRAND™ Discharge Tip For Automatic Burettes
Product Code. 11603199 Shop All Brand Products
Change view
Click to view available options
No. per Pack:
10 Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. per Pack Unit Size
11603199 10 Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11603199 Supplier BRAND™ Supplier No. 11505
  • ONLINE EXCLUSIVE PRICE
    107.00 EUR
    101.50 EUR / Pack of 10
    Save 5.50 EUR
    5% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Dr. Schilling pattern
  • Glass discharge tip
  • Dr. Schilling pattern
  • Includes silicone tubing
  • TRUSTED_SUSTAINABILITY
    Specifications
    Color Clear
    For Use With (Equipment) Automatic Burette
    Material Glass
    No. per Pack 10 Pk.
    Type Discharge Tip