Learn More
Buerkle™ Discharge Hose for Buerkle™ AccuOne/EnergyOne Barrel Pump
Product Code. 11332880 Shop All Bürkle Products
Change view
Click to view available options
Overall Height (Metric):
1.2 m
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Overall Height (Metric) Unit Size
11332880 1.2 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11332880 Supplier Buerkle™ Supplier No. 59003045
  • ONLINE EXCLUSIVE PRICE
    105.00 EUR
    103.60 EUR / Pack of 1
    Save 1.40 EUR
    1% Off
Estimated Shipment: 22-10-2026
Discharge hose 1.2m with nozzle for AccuOne/EnergyOne barrel pumps. Liquid-contacting parts constructed from PVC, PP, V2A, FKM
Discharge hose 1.2m with nozzle for AccuOne/EnergyOne barrel pumps. Liquid-contacting parts constructed from PVC, PP, V2A, FKM
TRUSTED_SUSTAINABILITY
Specifications
Includes Nozzle
Overall Height (Metric) 1.2 m
Type Discharge Hose