Learn More
Dipyrone, TRC
Product Code. 30135817
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135817 10 g 10g
17980885 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30135817 Supplier Toronto Research Chemicals Supplier No. TRCD49286010G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 68-89-3
Molecular Formula C13 H16 N3 O4 S . Na
Molecular Weight (g/mol) 333.34
Synonym Methanesulfonic acid, 1-[(2,3-dihydro-1,5-dimethyl-3-oxo-2-phenyl-1H-pyrazol-4-yl)methylamino]-, sodium salt (1:1), Dipyrone (6CI), Methanesulfonic acid, (antipyrinylmethylamino)-, sodium salt (8CI), Methanesulfonic acid, [(2,3-dihydro-1,5-dimethyl-3-oxo-2-phenyl-1H-pyrazol-4-yl)methylamino]-, sodium salt (9CI), ARPF, Algocalmin, Algopyrin, Anador, Analgin, Analgine, Andolor, Conmel, Diprofarn, Gifaril, Lisador, Metamizol, Metamizole, Metamizole sodium, Metamizole sodium salt, Metapyrin, Methamizole sodium, Methampyrone, Methylmelubrin, Metilon, Narone, Neo-melubrine, Noramidopyrine methanesulfonate sodium, Noramidopyrine methanesulfonate sodium salt, Noraminophenazone methanesulfonate sodium salt, Noraminophenazone sodium mesylate, Novalgetol, Novalgin, Novamidazophen, Novaminophenazone, Novaminsulfone, Novaminsulfone sodium, Novaminsulfonium, Novaminsulfonum, Noveltex, Optalgin, Pharmalgine, Pyralgine, Pyretin, Pyretin (pharmaceutical), Restriva, Sodium (antipyrinylmethylamino)methanesulfonate, Sodium 1-phenyl-2,3-dimethyl-5-pyrazolone-4-methylaminomethanesulfonate, Sodium metamizole, Sodium noramidopyrine methanesulfonate, Sodium novaminsulfonate, Sulpin, Sulpyrin, Sulpyrine
IUPAC Name sodium;[(1,5-dimethyl-3-oxo-2-phenylpyrazol-4-yl)-methylamino]methanesulfonate
SMILES [Na+].CN(CS(=O)(=O)[O-])C1=C(C)N(C)N(C1=O)c2ccccc2
Specifications
Quantity 10 g
InChI Formula InChI=1S/C13H17N3O4S.Na/c1-10-12(14(2)9-21(18,19)20)13(17)16(15(10)3)11-7-5-4-6-8-11;/h4-8H,9H2,1-3H3,(H,18,19,20);/q;+1/p-1
Formula Weight 333.0759
Chemical Name or Material Metamizol sodium