Learn More
Dipyridamole, TRC
Product Code. 30134969
Change view
Click to view available options
Quantity:
50 mg
500 mg
1 g
Unit Size:
1g
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134969 500 mg 500mg
30132473 1 g 1g
17911905 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134969 Supplier Toronto Research Chemicals Supplier No. TRCD492625500MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 58-32-2
Molecular Formula C24 H40 N8 O4
Molecular Weight (g/mol) 504.63
Synonym Dipyridamole, 2,2',2' ',2' ' '-[[4,8-Di(piperidin-1-yl)pyrimido[5,4-d]pyrimidine-2,6-diyl]dinitrilo]tetraethanol, Ethanol, 2,2',2' ',2' ' '-[(4,8-di-1-piperidinylpyrimido[5,4-d]pyrimidine-2,6-diyl)dinitrilo]tetrakis-, Ethanol, 2,2',2' ',2' ' '-[(4,8-dipiperidinopyrimido[5,4-d]pyrimidine-2,6-diyl)dinitrilo]tetra- (6CI,8CI), 2,2',2' ',2' ' '-[(4,8-Di-1-piperidinylpyrimido[5,4-d]pyrimidine-2,6-diyl)dinitrilo]tetrakis[ethanol], Pyrimido[5,4-d]pyrimidine, ethanol deriv., 2,2',2' ',2' ' '-((4,8-Di(piperidin-1-yl)pyrimido[5,4-d]pyrimidine-2,6-diyl)bis(azanetriyl))tetraethanol, 2,2',2' ',2' ' '-[[4,8-Dipiperidinopyrimido(5,4-d)pyrimidine-2,6-diyl]dinitrilo]tetraethanol, 2,6-Bis(diethanolamino)-4,8-dipiperidinopyrimido[5,4-d]pyrimidine, Anginal, Apricor, Cardioflux, Cardoxil, Cardoxin, Cleridium, Coribon, Coridil, Coronarine, Corosan, Coroxin, Curantyl, Dipyridamine, Dipyridamol, Dipyridan, Gulliostin, Kurantil, NSC 515776, NSC 619103, Natyl, Peridamol, Persantin, Persantine, Piroan, Prandiol, Protangix, RA 8, Stenocardil, Stimolcardio
IUPAC Name 2-[[2-[bis(2-hydroxyethyl)amino]-4,8-di(piperidin-1-yl)pyrimido[5,4-d]pyrimidin-6-yl]-(2-hydroxyethyl)amino]ethanol
SMILES OCCN(CCO)c1nc(N2CCCCC2)c3nc(nc(N4CCCCC4)c3n1)N(CCO)CCO
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C24H40N8O4/c33-15-11-31(12-16-34)23-26-20-19(21(27-23)29-7-3-1-4-8-29)25-24(32(13-17-35)14-18-36)28-22(20)30-9-5-2-6-10-30/h33-36H,1-18H2
Formula Weight 504.3173
Chemical Name or Material Dipyridamole