missing translation for 'onlineSavingsMsg'
Learn More

Dipyridamole, MedChemExpress

Product Code. 16488226
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488226 100 mg 100mg
16452096 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16488226 Supplier Medchem Express Supplier No. HYB0312
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Dipyridamole is an antiplatelet agent used in secondary prophylaxis against stroke.. Dipyridamole also can induce cancer cell-specific apoptosis as a phosphodiesterase (PDE) inhibitor..

Dipyridamole is an antiplatelet agent used in secondary prophylaxis against stroke.. Dipyridamole also can induce cancer cell-specific apoptosis as a phosphodiesterase (PDE) inhibitor..

Specifications

Chemical Name or Material Dipyridamole
Physical Form Solid
CAS 58-32-2
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C24H40N8O4
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 50 mg/mL (99.08 mM) ∣H2O : 0.67 mg/mL (1.33 mM; Need ultrasonic)
SMILES OCCN(CCO)C1=NC(N2CCCCC2)=C(N=C(N(CCO)CCO)N=C3N4CCCCC4)C3=N1
Molecular Weight (g/mol) 504.63
Formula Weight 504.63
Percent Purity 99.7%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.