Learn More
Dipropyl Sulfide, Sigma-Aldrich™
Specifications
Specifications
| CAS | 111-47-7 |
| Melting Point | -102°C |
| Density | 0.838 g/cm3 at 20°C |
| Boiling Point | 141°C to 142°C/1013 hPa |
| MDL Number | MFCD00009379 |
| Quantity | 25 mL |
| Synonym | Dipropyl sulfide, Dipropyl sulfide |
| Molecular Weight (g/mol) | 118.24 |
| Grade | Research |