Learn More
Diphenylmethane 4,4′-Diisocyanate, Sigma-Aldrich™
Product Code. 15871181
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15871181 100 mL 100mL
15816660 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15871181 Supplier Sigma Aldrich Supplier No. 8207970100
 Restricted Item
Estimated Shipment: 18-11-2026
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Mixture of di-and triisocyanates for synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 101-68-8
Specifications
Melting Point 10°C
Density 1.239 g/cm3 at 20°C
Boiling Point 196°C/7 hPa
MDL Number MFCD00036131
Quantity 100 mL
Synonym Diphenylmethane 4,4′-diisocyanate, Methylenediphenylene diisocyanate, Diphenylmethane 4,4′-diisocyanate
Molecular Weight (g/mol) 250.25
Grade Research