Learn More
Diphenylamine, Sigma-Aldrich™
Product Code. 15821011
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15821011 1 kg 1kg
15811011 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15821011 Supplier Sigma Aldrich Supplier No. 8205281000
 Restricted Item
Estimated Shipment: 19-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 122-39-4
Specifications
Melting Point 53°C to 54°C
Density 1.16 g/cm3 at 20°C
Boiling Point 302°C/1013 hPa
Molecular Formula (C6H5)2NH
MDL Number MFCD00003014
Quantity 1 kg
Synonym Diphenylamine, N-Phenylaniline, Diphenylamine
Molecular Weight (g/mol) 169.22
Grade Research