missing translation for 'onlineSavingsMsg'
Learn More

Diphenylamine-4-sulfonic Acid Barium Salt, Supelco™

Product Code. 15866830
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
missing translation for 'unitSize':
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
15866830 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15866830 missing translation for 'mfr' Supelco missing translation for 'supplierNo' 1002550005
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications

Redox indicator

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 300°C
CAS 6211-24-1
Quantity 5 g
MDL Number MFCD00007497
Synonym Diphenylamine-4-sulfonic acid barium salt, 4-Anilinobenzene sulfonic acid barium salt, Barium diphenylaminesulfonate, Diphenylamine-4-sulfonic acid barium salt
Molecular Weight (g/mol) 633.88
Grade Research
Chemical Name or Material Diphenylamine-4-Sulfonic Acid Barium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.