missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl Ether, Sigma-Aldrich™

Product Code. 15891301
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Unit Size:
1 L
100mL
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15891301 100 mL 100mL
15801311 1 L 1 L
15811311 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15891301 Supplier Sigma-Aldrich Supplier No. 8209780100
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-84-8

Specifications

Melting Point 24°C to 27°C
Density 1.07 g/cm3 at 20°C
Boiling Point 259°C/1013 hPa
Molecular Formula (C6H5)2O
MDL Number MFCD00003034
Quantity 100 mL
Synonym Diphenyl ether, Phenyl ether, Diphenyl oxide, Diphenyl ether
Molecular Weight (g/mol) 170.21
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.