missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenyl-d10 Ether, CDN
Specifications
Specifications
| Chemical Name or Material | Diphenyl ether-d10 |
| CAS | 93952-05-7 |
| Molecular Formula | C12 2H10 O |
| Quantity | 0.5 g |
| Synonym | Benzene-d5, 6,6'-oxybis- (9 CI), Oxydibenzene D10, 1,1'-Oxybis[benzene] D10, Biphenyl oxide D10, Diphenyl ether D10, Diphenyl oxide D10, Oxybisbenzene D10, Phenoxybenzene D10, Phenyl oxide D10 |
| InChI Formula | InChI=1 S/C12H10O/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h1-10 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(Oc2c([2 H])c([2 H])c([2 H])c([2 H])c2[2 H])c([2 H])c1[2 H] |
| IUPAC Name | 1,2,3,4,5-pentadeuterio-6-(2,3,4,5,6-pentadeuteriophenoxy)benzene |
| Molecular Weight (g/mol) | 180.27 |
| Formula Weight | 180.1359 g/mol |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?