Learn More
Thermo Scientific™ Dionex™ Specially Formulated PTFE Tubing
Product Code. 11352133 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
250 and 350 um ID
250 um ID; Low pressure
350 um ID; Low pressure
For Use With (Equipment):
Dionex UltiMate 3000 RSLC Nano System
Fused Silica Capillary Tubing
Unit Size:
Pack of 2
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description For Use With (Equipment) Unit Size
11352133 250 and 350 um ID Fused Silica Capillary Tubing Pack of 2
11322133 250 um ID; Low pressure Dionex UltiMate 3000 RSLC Nano System Pack of 5
11362143 350 um ID; Low pressure Fused Silica Capillary Tubing Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11352133 Supplier Thermo Scientific™ Supplier No. 160489
Estimated Shipment: 03-12-2026
Lägg i varukorgen
Lägg i varukorgen
Denna artikel kan inte returneras. Se returpolicy
Thermo Scientific™ Dionex Specially Formulated PTFE Tubing is used as connector for fused silica tubing (nano and cap LC). Choose fitting dimensions for OD of fused silica tubing used.
Thermo Scientific™ Dionex Specially Formulated PTFE Tubing is used as connector for fused silica tubing (nano and cap LC).
TRUSTED_SUSTAINABILITY
Specifikationer
Description 250 and 350 um ID
For Use With (Equipment) Fused Silica Capillary Tubing
Product Line nanoLC
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.