Learn More
Dinoprost, MedChemExpress
Product Code. 16498157
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498157 5 mg 5mg
16455448 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16498157 Supplier Medchem Express Supplier No. HY12956
MedChemExpress Dinoprost (Prostaglandin F2α) is an orally active, potent prostaglandin F (PGF) receptor (FP receptor) agonist. Dinoprost is a luteolytic hormone produced locally in the endometrial luminal epithelium and corpus luteum (CL). Dinoprost plays a key role in the onset and progression of labour.
Dinoprost (Prostaglandin F2α) is an orally active, potent prostaglandin F (PGF) receptor (FP receptor) agonist. Dinoprost is a luteolytic hormone produced locally in the endometrial luminal epithelium and corpus luteum (CL). Dinoprost plays a key role in the onset and progression of labour.
Specifications
Chemical Name or Material Dinoprost
Physical Form Oil
CAS 551-11-1
Color Brown
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C20H34O5
For Use With (Application) Metabolism-protein/nucleotide metabolism
Synonym Prostaglandin F2α PGF2α
Solubility Information DMSO : 100 mg/mL (282.10 mM; Need ultrasonic) ∣H2O : 100 mg/mL (282.10 mM; Need ultrasonic)
SMILES CCCCC[C@H](O)/C=C/[C@H]1[C@H](O)C[C@H](O)[C@@H]1C/C=C\CCCC(O)=O
Molecular Weight (g/mol) 354.48
Formula Weight 354.48
Percent Purity 99.06%
Grade Research