missing translation for 'onlineSavingsMsg'
Learn More

Dinitolmide, MedChemExpress

Product Code. 16484517
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16484517 50 mg 50mg
16448556 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16484517 Supplier Medchem Express Supplier No. HYB1004
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Dinitolmide (Zoalene), a fodder additive for poultry, has anti-coccidial effect. Dinitolmide can be used to prevent infections induced by Eimeria, such as Eimeria tenella, Eimeria necatrix, Eimeria brunette, and so on.

Dinitolmide (Zoalene), a fodder additive for poultry, has anti-coccidial effect. Dinitolmide can be used to prevent infections induced by Eimeria, such as Eimeria tenella, Eimeria necatrix, Eimeria brunette, and so on.

Specifications

Chemical Name or Material Dinitolmide
Physical Form Solid
CAS 148-01-6
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C8H7N3O5
For Use With (Application) COVID-19-immunoregulation
Synonym Zoalene
Solubility Information DMSO : ≥ 49 mg/mL (217.62 mM)
SMILES O=C(N)C1=CC([N+]([O-])=O)=CC([N+]([O-])=O)=C1C
Molecular Weight (g/mol) 225.16
Formula Weight 225.16
Percent Purity 99.48%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.