missing translation for 'onlineSavingsMsg'
Learn More

Diminazene Dihydrochloride, TRC

Product Code. 30138230
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138230 25 mg 25mg
18030620 10 mg 10mg
18060410 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138230 Supplier Toronto Research Chemicals Supplier No. TRCD47955025MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31384-83-5
Molecular Formula C14H17Cl2N7
Molecular Weight (g/mol) 354.24
Synonym 4,4'-(1-Triazene-1,3-diyl)bisbenzenecarboximidamide Dihydrochloride, 4,4'-(Diazoamino)dibenzamidine Dihydrochloride, Berenil Dihydrochloride;
SMILES NC(C1=CC=C(C=C1)N/N=N/C2=CC=C(C(N)=N)C=C2)=N

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C14H15N7.2ClH/c15-13(16)9-1-5-11(6-2-9)19-21-20-12-7-3-10(4-8-12)14(17)18;;/h1-8H,(H3,15,16)(H3,17,18)(H,19,20);2*1H
Formula Weight 281.1389
Percent Purity >95%
Chemical Name or Material Diminazene Dihydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.