missing translation for 'onlineSavingsMsg'
Learn More

Dimethylglyoxime, For ACS analysis, 99.49%, MP Biomedicals™

Product Code. 11432832 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11432832 500 g 500g
11412832 25 g 25g
11422832 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11432832 Supplier MP Biomedicals Supplier No. 0215251890

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 95-45-4
Molecular Formula C4H8N2O2
Molecular Weight (g/mol) 116.12
MDL Number MFCD00002117
InChI Key JGUQDUKBUKFFRO-GGWOSOGESA-N
Synonym biacetyl dioxime, dimethylglyoxime, diacetyldioxime, 2,3-butanedione dioxime, diacetyl dioxime, 2,3-diisonitrosobutane, biacetyl, dioxime, chugaev's reagent, nsc 9, unii-2971mft1ky
PubChem CID 5356010
IUPAC Name (E)-N-[(3E)-3-(hydroxyimino)butan-2-ylidene]hydroxylamine
SMILES C\C(=N/O)\C(\C)=N\O

Specifications

Color White
Assay Percent Range 0.9949
Quantity 500 g
Formula Weight 116.1
Physical Form Powder
Chemical Name or Material Dimethylglyoxime, For ACS analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.