Learn More
Dimethyle4-(4-methoxyphenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, 97%, Thermo Scientific™
Product Code. p-2703880
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10039291 1 g Amber Glass Bottle 1g
10374102 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10039291 Supplier Thermo Scientific Maybridge Supplier No. SEW02190DA
Chemical Identifiers
CAS 73257-47-3
Molecular Formula C18H21NO5
Molecular Weight (g/mol) 331.368
MDL Number MFCD00085027
InChI Key IAXDEFZXLVTHLU-UHFFFAOYSA-N
Synonym dimethyl 4-4-methoxyphenyl-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, 3,5-dimethyl 4-4-methoxyphenyl-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, dimethyl 4-4-methoxyphenyl-2,6-dimethyl-1,4-dihydro-3,5-pyridinedicarboxylate, maybridge1_007478, cbmicro_033161, 1,4-dihydro-2,6-dimethyl-4-4-methoxyphenyl-3,5-pyridinedicarboxylic acid dimethyl ester, dimethyl 1,4-dihydro-4-4-methoxyphenyl-2,6-dimethylpyridine-3,5-dicarboxylate, dimethyl 4-4-methoxyphenyl-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxy late, 3,5-pyridinedicarboxylic acid, 1,4-dihydro-4-4-methoxyphenyl-2,6-dimethyl-, dimethyl ester, 3,5-pyridinedicarboxylicacid, 1,4-dihydro-4-4-methoxyphenyl-2,6-dimethyl-, 3,5-dimethyl ester
PubChem CID 614332
IUPAC Name dimethyl 4-(4-methoxyphenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CC1=C(C(C(=C(N1)C)C(=O)OC)C2=CC=C(C=C2)OC)C(=O)OC
Specifications
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 331.37
Percent Purity 97%
Chemical Name or Material Dimethyl 4-(4-methoxyphenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate