Learn More
Dimethylarsinic Acid Sodium Salt Trihydrate, Sigma-Aldrich™
Product Code. 15844551
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15844551 250 g 250g
15824551 25 g 25g
15834551 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15844551 Supplier Sigma-Aldrich Supplier No. 8206700250
 Restricted Item
Estimated Shipment: 23-11-2026
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6131-99-3
Specifications
Melting Point 60°C
Quantity 250 g
MDL Number MFCD00149079
Synonym Dimethylarsinic acid sodium salt trihydrate, Dimethylarsinic acid sodium salt, Cacodylic acid sodium salt, Dimethylarsinic acid sodium salt trihydrate
Molecular Weight (g/mol) 214.03
Grade Ungraded
Chemical Name or Material Dimethylarsinic Acid Sodium Salt Trihydrate