missing translation for 'onlineSavingsMsg'
Learn More

Dimethylammonium Chloride, Sigma-Aldrich™

Product Code. 15896330
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
Unit Size:
1kg
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15896330 250 g 250g
15838910 5 g 5g
15848910 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15896330 Supplier Sigma Aldrich Supplier No. 8030520250
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-59-2

Specifications

Melting Point 160°C
Molecular Formula (CH3)2NH · HCl
MDL Number MFCD00012477
Quantity 250 g
Synonym Dimethylammonium chloride, Dimethylammonium chloride
Molecular Weight (g/mol) 81.54
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.