missing translation for 'onlineSavingsMsg'
Learn More

Dimethylamine, Sigma-Aldrich™

Product Code. 15896690
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15896690 2.5 L 2.5 L
15886690 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15896690 Supplier Sigma Aldrich Supplier No. 8220332500
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

40% Aqueous solution for synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-40-3

Specifications

Melting Point -38°C
Molecular Formula (CH3)2NH
MDL Number MFCD00008288
Density 0.89 g/cm3 at 20°C
Synonym Dimethylamine solution, Dimethylamine solution
Molecular Weight (g/mol) 45.08
Grade Reagent
Quantity 2.5 L
Chemical Name or Material Dimethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.