missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl terephthalate-2,3,5,6-d4, 98 atom % D, Thermo Scientific Chemicals

Product Code. 15449145
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15449145 250 mg 250mg
15457987 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15449145 Supplier Thermo Scientific Alfa Aesar Supplier No. H51104.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74079-01-9
Molecular Formula C10H10O4
Molecular Weight (g/mol) 198.21
MDL Number MFCD00182544
InChI Key WOZVHXUHUFLZGK-LNFUJOGGSA-N
Synonym dimethyl terephthalate-2,3,5,6-d4, 1,4-benzenedicarboxylic acid-d4 dimethyl ester, dimethyl p-phthalate-d4, dmt-d4, dimethyl terephthalate-d4, dimethyl p-benzenedicarboxylate-d4, dimethyl terephthalate-d4 ring-d4, methyl 4-carbomethoxy benzoate-d4, terephthalic acid-d4 dimethyl ester, dimethyl 1,4-benzenedicarboxylate-d4
PubChem CID 12241382
IUPAC Name dimethyl 2,3,5,6-tetradeuteriobenzene-1,4-dicarboxylate
SMILES COC(=O)C1=CC=C(C=C1)C(=O)OC

Specifications

Quantity 250 mg
Formula Weight 198.21
Percent Purity 95%
Assay 98 atom % D
Chemical Name or Material Dimethyl terephthalate-2,3,5,6-d4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.