Learn More
Dimethyl succinate, 98%
Product Code. p-7020564
Change view
Click to view available options
Quantity:
100 g
250 g
1000 g
Unit Size:
1000g
100g
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11364479 100 g 100g
11454264 250 g 250g
11444264 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11364479 Supplier Thermo Scientific Alfa Aesar Supplier No. A12565.22
Add to basket
CAS: 106-65-0 | C6H10O4 | 146.142 g/mol
Dimethyl succinate is used as a flavoring agent. It have wide range of industrial applications such as Functional fluids (open systems), Intermediates, Paint additives and coating additives, Pigments Solvents, Viscosity adjustors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dimethyl succinate is used as a flavoring agent. It have wide range of industrial applications such as Functional fluids (open systems), Intermediates, Paint additives and coating additives, Pigments Solvents, Viscosity adjustors.
Solubility
Soluble in water. (122,9 g/L at 20°C). soluble in acetone, ethanol, very soluble in ethyl ether.
Notes
Store at room temperature. Ensure good ventilation.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 106-65-0
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.142
MDL Number MFCD00008466
InChI Key MUXOBHXGJLMRAB-UHFFFAOYSA-N
Synonym dimethyl succinate, methyl succinate, dimethylsuccinate, butanedioic acid, dimethyl ester, succinic acid, dimethyl ester, dbe-4 dibasic ester, methyl butanedioate, succinic acid dimethyl ester, butanedioic acid, 1,4-dimethyl ester, fema no. 2396
PubChem CID 7820
IUPAC Name dimethyl butanedioate
SMILES COC(=O)CCC(=O)OC
Specifications
Melting Point 18°C to 19°C
Density 1.119
Boiling Point 196°C
Flash Point 85°C (185°F)
Refractive Index 1.419
Quantity 100 g
Beilstein 956776
Merck Index 14,8869
Solubility Information Soluble in water. (122,9g/L at 20°C). soluble in acetone,ethanol,very soluble in ethyl ether.
Formula Weight 146.14
Percent Purity ≥98%
Chemical Name or Material Dimethyl succinate