missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Dimethyl pimelimidate dihydrochloride has also been used to develop a technique for isolating Enterocytozoon bieneusi spores directly from human stool samples with a high degree of purity, which opens up new approaches for studying this parasite.
Specifications
Specifications
| CAS | 58537-94-3 |
| Melting Point | 122°C (decomposition) (literature) |
| Molecular Formula | C9H18N2O2 |
| MDL Number | MFCD00038942 |
| Quantity | 250 mg |
| Synonym | dimethyl pimelimidate dihydrochloride, pimelimidic acid dimethyl ester dihydrochloride, acmc-209tih, dimethyl pimelinediimidate dihydrochloride, dimethyl pimelinediimidate dihydro-chloride, dimethyl heptanebis imidate dihydrochloride, dimethyl pimelimidate dihydrochloride, powder, dimethyl pimelinediimidate paragraph signthornnieani, dimethyl pimelimidate dihydrochloride at |
| Solubility Information | Soluble in water (10mg/mL, clear) and methanol. |
| InChI Key | LRPQMNYCTSPGCX-UHFFFAOYSA-N |
| SMILES | COC(=N)CCCCCC(=N)OC |
| IUPAC Name | 1,7-dimethyl heptanediimidate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?