missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl pimelimidate, dihydrochloride, 98.9%, MP Biomedicals™

Product Code. 11496722 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496722 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11496722 Supplier MP Biomedicals Supplier No. 0215094505
  • ONLINE EXCLUSIVE PRICE
    314.00 EUR
    298.20 EUR / 5g
    Save 15.80 EUR
    5% Off
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 58537-94-3
Color White
Assay Percent Range 98.9%
Molecular Formula C9H18N2O2
MDL Number MFCD00038942
Quantity 5 g
Synonym dimethyl pimelimidate dihydrochloride, pimelimidic acid dimethyl ester dihydrochloride, acmc-209tih, dimethyl pimelinediimidate dihydrochloride, dimethyl pimelinediimidate dihydro-chloride, dimethyl heptanebis imidate dihydrochloride, dimethyl pimelimidate dihydrochloride, powder, dimethyl pimelinediimidate paragraph signthornnieani, dimethyl pimelimidate dihydrochloride at
InChI Key LRPQMNYCTSPGCX-UHFFFAOYSA-N
SMILES COC(=N)CCCCCC(=N)OC
IUPAC Name 1,7-dimethyl heptanediimidate
Molecular Weight (g/mol) 186.26
PubChem CID 11402688
Formula Weight 259.2
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl pimelimidate, dihydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.