Learn More
Dimethyl Phosphate, TRC
Product Code. 30136170
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136170 1 g 1g
30135835 250 mg 250mg
17923595 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30136170 Supplier Toronto Research Chemicals Supplier No. TRCD4769951G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 813-78-5
Molecular Formula C2 H7 O4 P
Molecular Weight (g/mol) 126.05
Synonym Methyl phosphate ((CH3O)2(HO)PO) (6CI), DMP, Dimethyl hydrogen phosphate, Dimethyl phosphate, NSC 2676, O,O-Dimethyl hydrogen phosphate, O,O-Dimethyl phosphate, Dimethylphosphate
IUPAC Name dimethyl hydrogen phosphate
SMILES COP(=O)(O)OC
Specifications
Quantity 1 g
InChI Formula InChI=1S/C2H7O4P/c1-5-7(3,4)6-2/h1-2H3,(H,3,4)
Formula Weight 126.0082
Percent Purity >95%
Chemical Name or Material Dimethyl phosphate
Grade HPLC